siRNA Details
Gene Symbol: Gm34517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132743.1 | - | 5 | 792 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB679762.1 | - | 4 | 346 | Chr. 19 - 43268935 - 43277655 [+] on Build GRCm38 |
