siRNA Details
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077468.1 | - | 1 | 1349 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV175131.2 | - | 1 | 303 | Chr. 1 - 44116772 - 44117750 [-] on Build GRCm38 |
