siRNA Details
Gene Symbol: 6330405D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018123.1 | - | 1 | 1128 | ||
AK161433.1 | - | 1 | 305 | |||
BB615983.1 | - | 1 | 305 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6330405D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018123.1 | - | 1 | 1128 | ||
AK161433.1 | - | 1 | 305 | |||
BB615983.1 | - | 1 | 305 |