siRNA Details
Gene Symbol: Gm15471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040412.1 | 5 | 609 | |||
GenBank mRNA | AK052792.1 | - | 4 | 1215 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB505891.2 | - | 2 | 640 | Chr. 3 - 103804597 - 103808438 [+] on Build GRCm38 |
