siRNA Details
Gene Symbol: Gm12530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040669.1 | 3 | 325 | |||
GenBank mRNA | AK052790.1 | - | 3 | 326 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661701.1 | - | 3 | 324 | Chr. 4 - 57172072 - 57175075 [+] on Build GRCm38 |
