siRNA Details
Gene Symbol: 9530036O11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053224.1 | - | 794 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB535728.2 | - | 6 | 596 | Chr. 5 - 28564358 - 28933995 [+] on Build GRCm38 |
