siRNA Details
Gene Symbol: 1700018A23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006078.1 | - | 1 | 63 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706191.1 | - | 1 | 63 | Chr. 6 - 126822377 - 126825177 [+] on Build GRCm38 |
