siRNA Details
Gene Symbol: 1700037C06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006618.1 | - | 1 | 644 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF452515.1 | - | 1 | 82 | Chr. 4 - 137116705 - 137117022 [-] on Build GRCm38 |
