siRNA Details
Gene Symbol: Gm26995 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146140.1 | - | 375 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY737654.1 | - | 4 | 375 | Chr. 5 - 117936387 - 117939520 [-] on Build GRCm38 |
