siRNA Details
Gene Symbol: A930003G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020804.1 | - | 1 | 253 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742115.1 | - | 1 | 253 | Chr. 7 - 12803546 - 12804202 [+] on Build GRCm38 |
