siRNA Details
Gene Symbol: A930003G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020804.1 | - | 1 | 768 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG802959.1 | - | 1 | 479 | Chr. 7 - 12803834 - 12804429 [+] on Build GRCm38 |
