siRNA Details
Gene Symbol: 1700074A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006945.1 | - | 1 | 230 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706969.1 | - | 1 | 228 | Chr. 2 - 181837417 - 181837798 [-] on Build GRCm38 |
