siRNA Details
Gene Symbol: 4930466I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015514.1 | - | 2 | 870 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715061.1 | - | 2 | 867 | Chr. 6 - 92013766 - 92016115 [-] on Build GRCm38 |
