siRNA Details
Gene Symbol: Gm2694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033430.1 | 4 | 511 | |||
GenBank mRNA | AK137304.1 | - | 4 | 512 | ||
AK144513.1 | - | 3 | 1284 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722377.1 | - | 4 | 512 | Chr. 8 - 87472812 - 87525523 [+] on Build GRCm38 |
