siRNA Details
Gene Symbol: Gm28382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031652.1 | - | 1 | 122 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB621301.1 | - | 1 | 118 | Chr. 1 - 91483027 - 91483584 [+] on Build GRCm38 |
