siRNA Details
Gene Symbol: Pard3bos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015421.1 | - | 483 | |||
AK076674.1 | - | 3 | 561 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV260171.2 | - | 3 | 453 | Chr. 1 - 61786747 - 61851433 [-] on Build GRCm38 |
