siRNA Details
Gene Symbol: Atxn2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155062.1 | - | 22 | 3453 | ||
AK158626.1 | - | 23 | 3022 | |||
AK168745.1 | - | 22 | 3123 | |||
AK219263.1 | - | 1 | 327 | |||
BC043451.1 | - | 17 | 2564 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC023938.1 | - | 1043 | Chr. - - [-] on Build |

More Information
Other Species Targeted

