siRNA Details
Gene Symbol: Atxn2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155062.1 | - | 22 | 3811 | ||
AK158626.1 | - | 23 | 3380 | |||
AK168745.1 | - | 22 | 3481 | |||
BC043451.1 | - | 17 | 2922 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC023938.1 | - | 1401 | Chr. - - [-] on Build |

More Information
Other Species Targeted

