siRNA Details
Gene Symbol: 4633401B06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014609.1 | - | 1 | 927 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK781410.1 | - | 1 | 518 | Chr. 3 - 121787539 - 121788161 [-] on Build GRCm38 |
