siRNA Details
Gene Symbol: 5033428I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034688.1 | - | 3 | 512 | ||
AK142377.1 | - | 1 | 587 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB626770.1 | - | 3 | 512 | Chr. 8 - 57332118 - 57333150 [+] on Build GRCm38 |
