siRNA Details
Gene Symbol: Gm41819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077070.1 | - | 1 | 208 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB573755.2 | - | 1 | 198 | Chr. 19 - 21992591 - 22020328 [+] on Build GRCm38 |
