siRNA Details
Gene Symbol: Gm26574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143393.1 | - | 2 | 529 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB666681.1 | - | 2 | 528 | Chr. 1 - 189834655 - 189836620 [-] on Build GRCm38 |
