siRNA Details
Gene Symbol: 4930442G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019609.1 | - | 2 | 244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714917.1 | - | 2 | 244 | Chr. 9 - 56931631 - 56932976 [+] on Build GRCm38 |
