siRNA Details
Gene Symbol: 2810427A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA667402.1 | - | 1 | 419 | ||
AK013168.1 | - | 1 | 527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712191.1 | - | 1 | 525 | Chr. 17 - 35920351 - 35920960 [+] on Build GRCm38 |
