siRNA Details
Gene Symbol: 4930439A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019601.1 | - | 1 | 15 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714892.1 | - | 1 | 15 | Chr. 1 - 37333533 - 37334172 [-] on Build GRCm38 |
