siRNA Details
Gene Symbol: 4930439D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019602.1 | - | 95 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714894.1 | - | 1 | 95 | Chr. 1 - 156939838 - 156940195 [+] on Build GRCm38 |
