siRNA Details
Gene Symbol: 5730402E23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019953.1 | - | 1 | 264 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717269.1 | - | 1 | 259 | Chr. 6 - 39810168 - 39810644 [-] on Build GRCm38 |
