siRNA Details
Gene Symbol: 4933401H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040540.1 | 1 | 18 | |||
GenBank mRNA | AK016602.1 | - | 1 | 19 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU936761.1 | - | 1 | 21 | Chr. 3 - 135833802 - 135840269 [-] on Build GRCm38 |
