siRNA Details
Gene Symbol: D730001M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021323.1 | - | 1 | 428 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661834.1 | - | 1 | 428 | Chr. 5 - 30206665 - 30207308 [+] on Build GRCm38 |
