siRNA Details
Gene Symbol: D730004N08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021329.1 | - | 1 | 627 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730753.1 | - | 1 | 624 | Chr. 2 - 110710343 - 110710977 [+] on Build GRCm38 |
