siRNA Details
Gene Symbol: 2810468A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013383.1 | - | 1 | 340 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712377.1 | - | 1 | 340 | Chr. 11 - 6547177 - 6547685 [+] on Build GRCm38 |
