siRNA Details
Gene Symbol: 1700094J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040580.1 | 5 | 920 | |||
GenBank mRNA | AK007067.1 | - | 5 | 919 |
Gene Symbol: Pcnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG088995.1 | - | 1 | 55 |
