siRNA Details
Gene Symbol: 6430584L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046179.1 | 4 | 881 | |||
GenBank mRNA | AK032533.1 | - | 4 | 882 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6430584L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046179.1 | 4 | 881 | |||
GenBank mRNA | AK032533.1 | - | 4 | 882 |