siRNA Details
Gene Symbol: Usp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086482.1 | - | 1 | 2197 | ||
BC082334.1 | - | 3 | 3641 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG277918.1 | - | 1 | 242 | Chr. 7 - 6769191 - 6769450 [+] on Build GRCm38 |
