siRNA Details
Gene Symbol: Svil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085972.1 | - | 1 | 102 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB662263.1 | - | 1 | 99 | Chr. 18 - 4994938 - 5002482 [+] on Build GRCm38 |
