siRNA Details
Gene Symbol: Bsph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020324.1 | - | 2, 3 | 122 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721134.1 | - | 2, 3 | 122 | Chr. 7 - 13554862 - 13571067 [-] on Build GRCm38 |
