siRNA Details
Gene Symbol: 4930474G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045398.1 | 4 | 276 | |||
GenBank mRNA | AK076809.1 | - | 7 | 600 | ||
AK136769.1 | - | 298 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB014047.2 | - | 7 | 629 | Chr. 18 - 28872218 - 28937986 [+] on Build GRCm38 |
