siRNA Details
Gene Symbol: 1700029M03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076808.1 | - | 12 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB013992.2 | - | 12 | Chr. 1 - 164960921 - 164962822 [+] on Build GRCm38 |
