siRNA Details
Gene Symbol: Gm14015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040637.1 | 3 | 3463 | |||
GenBank mRNA | AK033951.1 | - | 3 | 3465 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB076656.2 | - | 1 | 196 | Chr. 2 - 106463823 - 106464504 [-] on Build GRCm38 |
