siRNA Details
Gene Symbol: 4931408D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040298.1 | 2 | 366 | |||
NR_040299.1 | 2 | 366 | ||||
GenBank mRNA | AK016444.1 | - | 2 | 161 | ||
AK040817.1 | - | 2 | 288 | |||
AK168860.1 | - | 2 | 359 |
