siRNA Details
Gene Symbol: 1700008A23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005751.1 | - | 1 | 15 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA465510.1 | - | 1 | 75 | Chr. 9 - 99322769 - 99323496 [+] on Build GRCm38 |
