siRNA Details
Gene Symbol: Gm6936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045001.1 | 4 | 1241 | |||
GenBank mRNA | AK047639.1 | - | 4 | 1242 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm6936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045001.1 | 4 | 1241 | |||
GenBank mRNA | AK047639.1 | - | 4 | 1242 |