siRNA Details
Gene Symbol: BC049265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158865.1 | - | 773 | |||
BC030475.1 | - | 675 | ||||
BC049265.1 | - | 664 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI739087.1 | - | 1 | 664 | Chr. 7 - 108950766 - 108951553 [+] on Build GRCm38 |
