siRNA Details
Gene Symbol: Gm19626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044311.1 | - | 589 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB642240.1 | - | 1 | 589 | Chr. 17 - 53636130 - 53636803 [-] on Build GRCm38 |
