siRNA Details
Gene Symbol: 4930507D10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110449.1 | 2 | 239 | |||
GenBank mRNA | AK015727.1 | - | 2 | 240 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715269.1 | - | 2 | 240 | Chr. 11 - 80850633 - 80858300 [-] on Build GRCm38 |
