siRNA Details
Gene Symbol: 2310047D07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009869.1 | - | 2 | 367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709471.1 | - | 2 | 367 | Chr. 5 - 149237337 - 149237966 [+] on Build GRCm38 |
