siRNA Details
Gene Symbol: 1700125C05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007275.1 | - | 1 | 79 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707373.1 | - | 1 | 77 | Chr. 13 - 112800379 - 112800723 [-] on Build GRCm38 |
