siRNA Details
Gene Symbol: 1700124M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007272.1 | - | 2 | 277 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707371.1 | - | 2 | 277 | Chr. 10 - 19086527 - 19087178 [-] on Build GRCm38 |
