siRNA Details
Gene Symbol: Pcsk2os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040625.1 | 3 | 1622 | |||
GenBank mRNA | AK034766.1 | - | 3 | 1622 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA387407.1 | - | 3 | 379 | Chr. 2 - 143749164 - 143795965 [-] on Build GRCm38 |
