siRNA Details
Gene Symbol: Gm26787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007186.1 | - | 4 | 390 | ||
AK018988.1 | - | 4 | 369 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707377.1 | - | 4 | 369 | Chr. 9 - 36767910 - 36782677 [+] on Build GRCm38 |
